Få mere at vide
Pirenzepine Dihydrochloride Monohydrate, Mikromol™
Beskrivelse
Pirenzepine Dihydrochloride Monohydrate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Pirenzepine Dihydrochloride Monohydrate |
| CAS | 2242504-48-7 |
| Analyt- eller komponentnavne | Pirenzepine Dihydrochloride Monohydrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H21 N5 O2 . 2 Cl H . H2 O |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Pirenzepine dihydrochloride monohydrate, 11-[(4-Methylpiperazin-1-yl)acetyl]-5,11-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one dihydrochloride monohydrate |
| InChI formel | InChI=1S/C19H21N5O2.2ClH.H2O/c1-22-9-11-23(12-10-22)13-17(25)24-16-7-3-2-5-14(16)19(26)21-15-6-4-8-20-18(15)24;;;/h2-8H,9-13H2,1H3,(H,21,26);2*1H;1H2 |
| SMIL | O.Cl.Cl.CN1CCN(CC(=O)N2c3ccccc3C(=O)Nc4cccnc24)CC1 |
| IUPAC navn | 11-[2-(4-methylpiperazin-1-yl)acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one;hydrate;dihydrochloride |
| Vis mere |