Få mere at vide
Pirbuterol Acetate, TRC
Artikelnummer. 30416157
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30416157 50 mg
30416093 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30416157 Leverandør Toronto Research Chemicals Leverandørnr. TRCP50249550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 65652-44-0
Molekylær formel C12 H20 N2 O3 . C2 H4 O2
Molekylvægt (g/mol) 300.35
Synonym 2,6-Pyridinedimethanol, α6-[[(1,1-dimethylethyl)amino]methyl]-3-hydroxy-, acetate (1:1), 2,6-Pyridinedimethanol, α6-[[(1,1-dimethylethyl)amino]methyl]-3-hydroxy-, monoacetate (salt) (9CI), CP 24314-14, Maxair, Pirbuterol acetate, Spirolair
IUPAC navn acetic acid;6-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)pyridin-3-ol
SMIL CC(=O)O.CC(C)(C)NCC(O)c1ccc(O)c(CO)n1
Tekniske data
Kemisk navn eller materiale Pirbuterol Acetate
Mængde 50 mg
InChI formel InChI=1S/C12H20N2O3.C2H4O2/c1-12(2,3)13-6-11(17)8-4-5-10(16)9(7-15)14-8;1-2(3)4/h4-5,11,13,15-17H,6-7H2,1-3H3;1H3,(H,3,4)
Formel vægt 300.1685