missing translation for 'onlineSavingsMsg'
Få mere at vide

Piracetam, TRC

Artikelnummer. 30011193
Change view
Click to view available options
Mængde:
50 mg
5 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30011193 50 mg
30012107 50 g
17949724 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011193 Supplier Toronto Research Chemicals Supplier No. TRCP50080050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7491-74-9
Molekylær formel C6 H10 N2 O2
Molekylvægt (g/mol) 142.16
Synonym Piracetam, (2-Oxopyrrolidino)acetamide, 2-(2-Oxopyrrolidin-1-yl)acetamide, 2-Ketopyrrolidine-1-acetamide, 2-Oxo-1-pyrrolidineacetamide, 2-Oxo-1-pyrrolidinylacetamide, 2-Pyrrolidinoneacetamide, 2-Pyrrolidoneacetamide, Avigilen, Axonyl, Cerebroforte, Cl 781, Encetrop, Euvifor, Gabacet, Genogris, Geram, Myocalm, Nootobril, Nootobryl, Nootron, Nootrop, Nootropil, Nootropyl, Normabrain, Norzetam, Piramem, Pirazetam, Pirroxil, Prima, Prima (pharmaceutical), Pyracetam, Pyramem, UCB 6215
IUPAC navn 2-(2-oxopyrrolidin-1-yl)acetamide
SMIL NC(=O)CN1CCCC1=O

Specifications

Kemisk navn eller materiale Piracetam
Mængde 50 mg
InChI formel InChI=1S/C6H10N2O2/c7-5(9)4-8-3-1-2-6(8)10/h1-4H2,(H2,7,9)
Formel vægt 142.0742
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.