missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Piracetam EP Urenhed D, Mikromol™
Description
Piracetam EP Urenhed D
Specifications
Specifications
| Kemisk navn eller materiale | (2-Oxopyrrolidin-1-yl)acetic Acid |
| CAS | 53934-76-2 |
| Analyt- eller komponentnavne | (2-Oxopyrrolidin-1-yl)acetic Acid |
| Forsendelsestilstand | Stuetemperatur |
| Molekylær formel | C6 H9 N O3 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | (2-Oxopyrrolidin-1-yl)acetic Acid, Piracetam Imp. D (EP), 2-Oxo-1-pyrrolidineacetic acid, (2-Oxopyrrolidin-1-yl)acetic acid, 2-Oxo-1-pyrrolidineacetic acid, 2-Pyrrolidinon-1-ylacetic acid, 2-Pyrrolidinoneacetic acid, Piracetam Impurity D |
| InChI formel | InChI=1S/C6H9NO3/c8-5-2-1-3-7(5)4-6(9)10/h1-4H2,(H,9,10) |
| SMIL | OC(=O)CN1CCCC1=O |
| IUPAC navn | 2-(2-oxopyrrolidin-1-yl)eddikesyre |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?