Få mere at vide
Piracetam-d8, TRC
Artikelnummer. 30410838
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30410838 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410838 Leverandør Toronto Research Chemicals Leverandørnr. TRCP50080210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Piracetam-d8
CAS 1329799-64-5
Molekylær formel C6 D8 H2 N2 O2
Mængde 10 mg
Synonym 2,2-Dideuterio-2-(2,2,3,3,4,4-hexadeuterio-5-oxopyrrolidin-1-yl)acetamide, 2-Oxo-1-pyrrolidineacetamide-α,α,3,3,4,4,5,5-d8, 2-(2-Oxopyrrolidin-1-yl-3,3,4,4,5,5-d6)acetamide-2,2-d2, Piracetam-d8, Piracetam-D8
InChI formel InChI=1S/C6H10N2O2/c7-5(9)4-8-3-1-2-6(8)10/h1-4H2,(H2,7,9)/i1D2,2D2,3D2,4D2
SMIL [2H]C([2H])(N1C(=O)C([2H])([2H])C([2H])([2H])C1([2H])[2H])C(=O)N
IUPAC navn 2,2-dideuterio-2-(2,2,3,3,4,4-hexadeuterio-5-oxopyrrolidin-1-yl)acetamide
Molekylvægt (g/mol) 150.205
Formel vægt 150.124