Få mere at vide
Piracetam-d8, CDN
Artikelnummer. 30325630
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30325630 0.005 g
30323111 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325630 Leverandør CDN Leverandørnr. CDND73200.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Piracetam-d8
CAS 1329799-64-5
Molekylær formel C6 D8 H2 N2 O2
Mængde 0.005 g
Synonym 2,2-Dideuterio-2-(2,2,3,3,4,4-hexadeuterio-5-oxopyrrolidin-1-yl)acetamide, 2-Oxo-1-pyrrolidineacetamide-α,α,3,3,4,4,5,5-d8, 2-(2-Oxopyrrolidin-1-yl-3,3,4,4,5,5-d6)acetamide-2,2-d2, Piracetam-d8, Piracetam-D8
InChI formel InChI=1 S/C6H10N2O2/c7-5(9)4-8-3-1-2-6(8)10/h1-4H2,(H2,7,9)/i1D2,2D2,3D2,4D2
SMIL [2 H]C([2 H])(N1C(=O)C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H])C(=O)N
IUPAC navn 2,2-dideuterio-2-(2,2,3,3,4,4-hexadeuterio-5-oxopyrrolidin-1-yl)acetamide
Molekylvægt (g/mol) 150.205
Formel vægt 150.124 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity