Få mere at vide
Piprinhydrinate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Piprinhydrinate |
| CAS | 606-90-6 |
| Analyt- eller komponentnavne | Piprinhydrinate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H23 N O . C7 H7 Cl N4 O2 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Piprinhydrinate, 8-Chloro-3,9-dihydro-1,3-dimethyl-1H-purine-2,6-dione compd. with 4-(diphenylmethoxy)-1-methylpiperidine (1:1), 8-Chloro-3,7-dihydro-1,3-dimethyl-1H-purine-2,6-dione compd. with 4-(diphenylmethoxy)-1-methylpiperidine (1:1), 8-Chlorotheophylline compd. with 4-(diphenylmethoxy)-1-methylpiperidine (1:1), 4-(Diphenylmethoxy)-1-methylpiperidine compd. with 8-chloro-3,7-dihydro-1,3-dimethyl-1H-purine-2,6-dione (1:1), 4-(Diphenylmethoxy)-1-methylpiperidine compd. with 8-chlorotheophylline (1:1), 8-Chlorotheophylline 4-(diphenylmethoxy)-1-methylpiperidine salt, Diphenylpyraline 8-chlorotheophyllinate, Diphenylpyraline theoclate, Kolton, Mepedyl, N-Methylpiperidyl 4-benzhydryl ether 8-chlorotheophyllinate |
| InChI formel | InChI=1S/C19H23NO.C7H7ClN4O2/c1-20-14-12-18(13-15-20)21-19(16-8-4-2-5-9-16)17-10-6-3-7-11-17;1-11-4-3(9-6(8)10-4)5(13)12(2)7(11)14/h2-11,18-19H,12-15H2,1H3;1-2H3,(H,9,10) |
| Løsningstype | Neat |
| SMIL | CN1CCC(CC1)OC(c2ccccc2)c3ccccc3.CN4C(=O)N(C)c5nc(Cl)[nH]c5C4=O |
| Vis mere |