Learn More
Hamilton™ 300μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips
Essential for precise pipetting. Hamilton™ 300μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips provide enhanced pipetting safety and reliability with the CO-RE PerfectTouch system.
Brand: Hamilton™ 235902
Additional Details : Weight : 6.02100kg
Description
- 59.9mm tip length
- Color code label: yellow
- Usable with 1000μL pipetting channels/CO-RE 96 Probe Head
- Pressure-based Liquid Level Detection (pLLD) and capacitive LLD (cLLD) with conductive (black) version
- Process safety offered by Monitored Air Displacement and Total Aspiration and Dispense Monitoring
Specifications
300 μL | |
Non-sterile | |
Yellow |
Non-Filtered | |
Black | |
Single-use |