Learn More
Melford PIPES, Disodium Salt
Product Code. 30018705
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity Unit Size
30018705 100 g 100g
1 options
Product Code. 30018705 Supplier Melford Supplier No. P40180100.0
 Restricted Item
Forventet forsendelse: 11-12-2026
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
pKa (25°C): 6.8 The working pH range in aqueous solution is 6.5-7.5.
Specifications
Chemical Name or Material PIPES, Disodium Salt
CAS 76836-02-7
Quantity 100 g
Molecular Formula C8H16N2Na2O6S2