Få mere at vide
Piperonyl Butoxide-d9, TRC
Artikelnummer. 30417269
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30417269 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30417269 Leverandør Toronto Research Chemicals Leverandørnr. TRCP49020225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Piperonylbutoxide butyl-d9
CAS 1329834-53-8
Renhedsgrad noter HPLC
Molekylær formel C19 2H9 H21 O5
Mængde 25 mg
Synonym Piperonyl Butoxide D9 (butyl D9), 5-[2-[2-(1,1,2,2,3,3,4,4,4-Nonadeuteriobutoxy)ethoxy]ethoxymethyl]-6-propyl-1,3-benzodioxole, Piperonylbutoxide D9 (butyl D9), 5-[2-[2-(1,1,2,2,3,3,4,4,4-Nonadeuteriobutoxy)ethoxy]ethoxymethyl]-6-propyl-1,3-benzodioxole, Piperonylbutoxide D9 (butyl-D9)
InChI formel InChI=1S/C19H30O5/c1-3-5-7-20-8-9-21-10-11-22-14-17-13-19-18(23-15-24-19)12-16(17)6-4-2/h12-13H,3-11,14-15H2,1-2H3/i1D3,3D2,5D2,7D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])OCCOCCOCc1cc2OCOc2cc1CCC
IUPAC navn 5-[2-[2-(1,1,2,2,3,3,4,4,4-nonadeuteriobutoxy)ethoxy]ethoxymethyl]-6-propyl-1,3-benzodioxole
Molekylvægt (g/mol) 347.49
Formel vægt 347.2658
Procent renhed >95