Få mere at vide
Piperidolate Hydrochloride, Mikromol™
Beskrivelse
Piperidolate Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Piperidolate Hydrochloride |
| CAS | 129-77-1 |
| Analyt- eller komponentnavne | Piperidolate Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C21 H25 N O2 . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Acetic acid, diphenyl-, 1-ethyl-3-piperidyl ester hydrochloride (6CI,7CI,8CI), 3-Piperidinol, 1-ethyl-, diphenylacetate (ester), hydrochloride (8CI), Crapinon, Dactil hydrochloride, NSC 4349, Piperidolate hydrochloride, Benzeneacetic acid, alpha-phenyl-, 1-ethyl-3-piperidinyl ester, hydrochloride (1:1), Benzeneacetic acid, alpha-phenyl-, 1-ethyl-3-piperidinyl ester, hydrochloride (9CI) |
| InChI formel | InChI=1S/C21H25NO2.ClH/c1-2-22-15-9-14-19(16-22)24-21(23)20(17-10-5-3-6-11-17)18-12-7-4-8-13-18;/h3-8,10-13,19-20H,2,9,14-16H2,1H3;1H |
| SMIL | Cl.CCN1CCCC(C1)OC(=O)C(c2ccccc2)c3ccccc3 |
| IUPAC navn | (1-ethylpiperidin-3-yl) 2,2-diphenylacetate;hydrochloride |
| Vis mere |