Få mere at vide
Piperidione, LoGiCal
Artikelnummer. 30460788
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30460788 5 mg
1 options
Artikelnummer. 30460788 Leverandør LoGiCal Leverandørnr. LGCFOR1672.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Piperidione
CAS 77-03-2
Analyt- eller komponentnavne Piperidione
Forsendelsestilstand Room Temperature
Molekylær formel C9 H15 N O2
Certificeringer/compliance ISO 9001
Synonym 3,3-Diethyl-2,4-dioxopiperidine, 3,3-Diethylpiperidine-2,4-dione, Ascron, Dihyprylon, Dihyprylone, NSC 75628, Nu 1510, Piperidion, Piperidione, Sedulon, Tusseval, 2,4-Piperidinedione, 3,3-diethyl-
InChI formel InChI=1S/C9H15NO2/c1-3-9(4-2)7(11)5-6-10-8(9)12/h3-6H2,1-2H3,(H,10,12)
Løsningstype Neat
SMIL CCC1(CC)C(=O)CCNC1=O
IUPAC navn 3,3-diethylpiperidine-2,4-dione
Industry Type Clinical Testing
Molekylvægt (g/mol) 169.22
Mængde 5 mg
Formel vægt 169.1103 g/mol