Learn More
Piperidine-2,6-dione, 98%, BLD Pharm™
Product Code. 31032508
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
1 kg
Unit Size:
1g
1kg
5g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31032508 1 g 1g
31014315 100 g -
31017830 1 kg 1kg
31017094 10 g -
31030039 500 g -
31023157 5 g 5g
31032117 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 31032508 Supplier BLD Pharm Supplier No. BD1321231g
 Restricted Item
Forventet forsendelse: 27-11-2026
Specifications
CAS 1121-89-7
Molecular Formula C5H7NO2
MDL Number MFCD00006670
Quantity 1 g
SMILES O=C(CCC1)NC1=O
Molecular Weight (g/mol) 113.11
Percent Purity 98%
Chemical Name or Material Piperidine-2,6-dione