Få mere at vide
Piperazinedione-carbonyl D-Phenylglycyl-glycine, TRC
Artikelnummer. 30419431
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30419431 25 mg
1 options
Artikelnummer. 30419431 Leverandør Toronto Research Chemicals Leverandørnr. TRCP48016525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Piperazinedione-carbonyl D-Phenylglycyl-glycine
CAS 2170771-47-6
Molekylær formel C17H20N4O6
Mængde 25 mg
Synonym Piperazinedione-carbonyl D-phenylglycyl-glycine, [(2R)-2-(4-Ethyl-2,3-dioxopiperazine-1-carboxamido)-2-phenylacetamido]acetic acid
InChI formel InChI=1S/C17H20N4O6/c1-2-20-8-9-21(16(26)15(20)25)17(27)19-13(11-6-4-3-5-7-11)14(24)18-10-12(22)23/h3-7,13H,2,8-10H2,1H3,(H,18,24)(H,19,27)(H,22,23)/t13-/m1/s1
SMIL CCN1CCN(C(=O)N[C@@H](C(=O)NCC(=O)O)c2ccccc2)C(=O)C1=O
IUPAC navn 2-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetyl]amino]acetic acid
Molekylvægt (g/mol) 376.36
Formel vægt 376.1383