Få mere at vide
Piperazine Phosphate Monohydrate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Piperazine Phosphate |
| CAS | 14538-56-8 |
| Molekylær formel | C4 H10 N2 . H3 O4 P |
| Mængde | 500 mg |
| Synonym | Anthalazine Phosphate, Antipar, Parazine, Pincets, Pinrou, Pinsirup, Piperazate, Piperazine Monophosphate, Piperazine Phosphate |
| InChI formel | InChI=1S/C4H10N2.H3O4P/c1-2-6-4-3-5-1;1-5(2,3)4/h5-6H,1-4H2;(H3,1,2,3,4) |
| SMIL | OP(=O)(O)O.C1CNCCN1 |
| IUPAC navn | phosphoric acid;piperazine |
| Molekylvægt (g/mol) | 184.13 |
| Formel vægt | 184.0613 |