Få mere at vide
Piperazine-N,N'-bis(2-ethanesulfonic Acid)-d18, CDN
Artikelnummer. 30314999
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30314999 0.1 g 0.1g
30328704 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314999 Leverandør CDN Leverandørnr. CDND61230.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Piperazinediethanesulfonic-D18 Acid
CAS 352534-95-3
Molekylær formel C8 D18 N2 O6 S2
Mængde 0.1 g
Synonym 1,4-Piperazine-2,2,3,3,5,5,6,6-d8-diethane-α,α,β,β-d4-sulfonic acid-d (9 CI), 1,4-Piperazinediethanesulfonic-D18 acid, 1,4-Piperazinediethanesulfonic-d18 acid, 1,4-Piperazinebis(ethanesulfonic acid)-d18, Perdeutero-2,2'-piperazine-1,4-diyl-bis-ethanesulfonic acid, Piperazine-N,N'-bis(2-ethanesulfonic acid)-d18, Piperazine-N,N'-bis(2-ethanesulfonic acid)-d18, Piperazine-N,N'-bis(2-ethanesulfonic acid)-d18
InChI formel InChI=1 S/C8H18N2O6S2/c11-17(12,13)7-5-9-1-2-10(4-3-9)6-8-18(14,15)16/h1-8H2,(H,11,12,13)(H,14,15,16)/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2/hD2
SMIL [2 H]OS(=O)(=O)C([2 H])([2 H])C([2 H])([2 H])N1C([2 H])([2 H])C([2 H])([2 H])N(C([2 H])([2 H])C([2 H])([2 H])S(=O)(=O)O[2 H])C([2 H])([2 H])C1([2 H])[2 H]
IUPAC navn deuterio 1,1,2,2-tetradeuterio-2-[2,2,3,3,5,5,6,6-octadeuterio-4-(1,1,2,2-tetradeuterio-2-deuteriooxysulfonylethyl)piperazin-1-yl]ethanesulfonate
Molekylvægt (g/mol) 320.479
Formel vægt 320.174 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity