missing translation for 'onlineSavingsMsg'
Få mere at vide

Piperazine-d8 Trifluoroacetic Acid Salt, TRC

Artikelnummer. 30427669
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427669 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427669 Leverandør Toronto Research Chemicals Leverandørnr. TRCP480103100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Piperazine-d8 Trifluoroacetic Acid Salt
CAS 1246815-61-1
Molekylær formel C8H4D8F6N2O4
Mængde 100 mg
Synonym Piperazine-2,2,3,3,5,5,6,6-d8 Trifluoroacetic Acid Salt, Ascarex D-d8 Trifluoroacetic Acid Salt, Diethylenediamine-d8 Trifluoroacetic Acid Salt, Dihydro Pip Wormer-d8 Trifluoroacetic Acid Salt, Dowzene DHC-d8 Trifluoroacetic Acid Salt, Piperazinium-d8 Trifluoroacetic Acid Salt
SMIL [2H]C1([2H])NC([2H])([2H])C([2H])([2H])NC1([2H])[2H].OC(=O)C(F)(F)F
Molekylvægt (g/mol) 322.23
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.