Få mere at vide
Piperazine-3,3,5,5-d4-N-t-BOC, CDN
Artikelnummer. 30327840
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327840 0.25 g
30326273 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327840 Leverandør CDN Leverandørnr. CDND74670.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Boc-piperazine-3,3,5,5-D4
CAS 1126621-87-1
Molekylær formel C9 D4 H14 N2 O2
Mængde 0.25 g
Synonym 1-Piperazine-3,3,5,5-d4-carboxylic acid, 1,1-dimethylethyl ester (ACI), 1,1-Dimethylethyl 1-piperazine-3,3,5,5-d4-carboxylate (ACI), N-Boc-piperazine-d4, Piperazine-3,3,5,5-d4-N-t-BOC
InChI formel InChI=1 S/C9H18N2O2/c1-9(2,3)13-8(12)11-6-4-10-5-7-11/h10H,4-7H2,1-3H3/i4D2,5D2
SMIL [2 H]C1([2 H])CN(CC([2 H])([2 H])N1)C(=O)OC(C)(C)C
IUPAC navn tert-butyl 3,3,5,5-tetradeuteriopiperazine-1-carboxylate
Molekylvægt (g/mol) 190.2761
Formel vægt 190.1619 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity