Få mere at vide
Piperazine-2,2,3,3,5,5,6,6-d8, TRC
Artikelnummer. 30420429
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30420429 50 mg
30415576 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30420429 Leverandør Toronto Research Chemicals Leverandørnr. TRCP47990250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 134628-42-5
Molekylær formel C4 D8 H2 N2
Molekylvægt (g/mol) 94.19
Synonym Piperazine-2,2,3,3,5,5,6,6-d8 (9CI, ACI), 2,2,3,3,5,5,6,6-Octadeuteriopiperazine, Piperazine-d8, Piperazine-D8
IUPAC navn 2,2,3,3,5,5,6,6-octadeuteriopiperazine
SMIL [2H]C1([2H])NC([2H])([2H])C([2H])([2H])NC1([2H])[2H]
Tekniske data
Kemisk navn eller materiale Piperazine-2,2,3,3,5,5,6,6-d8
Mængde 50 mg
InChI formel InChI=1S/C4H10N2/c1-2-6-4-3-5-1/h5-6H,1-4H2/i1D2,2D2,3D2,4D2
Formel vægt 94.1346