Få mere at vide
Piperazine-2,2,3,3,5,5,6,6-d8, CDN
Artikelnummer. 30316540
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30316540 0.5 g 0.5g
30318119 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316540 Leverandør CDN Leverandørnr. CDND62830.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Piperazine-2,2,3,3,5,5,6,6-d8
CAS 134628-42-5
Molekylær formel C4 D8 H2 N2
Mængde 0.5 g
Synonym Piperazine-2,2,3,3,5,5,6,6-d8 (9 CI, ACI), 2,2,3,3,5,5,6,6-Octadeuteriopiperazine, Piperazine-d8, Piperazine-D8
InChI formel InChI=1 S/C4H10N2/c1-2-6-4-3-5-1/h5-6 H,1-4H2/i1D2,2D2,3D2,4D2
SMIL [2 H]C1([2 H])NC([2 H])([2 H])C([2 H])([2 H])NC1([2 H])[2 H]
IUPAC navn 2,2,3,3,5,5,6,6-octadeuteriopiperazine
Molekylvægt (g/mol) 94.19
Formel vægt 94.1346 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity