Få mere at vide
Piperaquine Tetraphosphate, TRC
Artikelnummer. 30138344
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138344 250 mg
30007962 1 g
30003707 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138344 Leverandør Toronto Research Chemicals Leverandørnr. TRCP480050250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 911061-10-4
Molekylær formel C29H32Cl2N6•4H3PO4
Molekylvægt (g/mol) 927.49
Synonym Quinoline, 4,4'-(1,3-propanediyldi-4,1-piperazinediyl)bis[7-chloro-, phosphate (1:4) (9CI, ACI), Piperaquine tetraphosphate, 4,4'-(1,3-Propanediyldi-4,1-piperazinediyl)bis[7-chloroquinoline] tetraphosphate
IUPAC navn 7-chloro-4-[4-[3-[4-(7-chloroquinolin-4-yl)piperazin-1-yl]propyl]piperazin-1-yl]quinoline;phosphoric acid
SMIL OP(=O)(O)O.OP(=O)(O)O.OP(=O)(O)O.OP(=O)(O)O.Clc1ccc2c(ccnc2c1)N3CCN(CCCN4CCN(CC4)c5ccnc6cc(Cl)ccc56)CC3
Tekniske data
Kemisk navn eller materiale Piperaquine Tetraphosphate
Mængde 250 mg
InChI formel InChI=1S/C29H32Cl2N6.4H3O4P/c30-22-2-4-24-26(20-22)32-8-6-28(24)36-16-12-34(13-17-36)10-1-11-35-14-18-37(19-15-35)29-7-9-33-27-21-23(31)3-5-25(27)29;4*1-5(2,3)4/h2-9,20-21H,1,10-19H2;4*(H3,1,2,3,4)
Formel vægt 926.1141