Få mere at vide
Piperaquine-d6 Tetraphosphate, TRC
Artikelnummer. 30414725
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30414725 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414725 Leverandør Toronto Research Chemicals Leverandørnr. TRCP48005210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Piperaquine-d6 Tetraphosphate
CAS 1261394-71-1
Molekylær formel C29H26D6Cl2N6•4H3PO4
Mængde 10 mg
Synonym 4,4'-(1,3-Propanediyldi-4,1-piperazinediyl)bis[7-chloroquinoline]-d6 Tetraphosphate
InChI formel InChI=1S/C29H32Cl2N6/c30-22-2-4-24-26(20-22)32-8-6-28(24)36-16-12-34(13-17-36)10-1-11-35-14-18-37(19-15-35)29-7-9-33-27-21-23(31)3-5-25(27)29/h2-9,20-21H,1,10-19H2
SMIL ClC1=CC2=C(C(N3CCN(C([2H])(C([2H])(C([2H])(N4CCN(CC4)C5=C6C=CC(Cl)=CC6=NC=C5)[2H])[2H])[2H])CC3)=CC=N2)C=C1.OP(O)(O)=O.OP(O)(O)=O.OP(O)(O)=O.OP(O)(O)=O
Molekylvægt (g/mol) 933.53