Få mere at vide
Pipendoxifene, TRC
Artikelnummer. 30424372
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424372 10 mg
30422959 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30424372 Leverandør Toronto Research Chemicals Leverandørnr. TRCP47985010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 198480-55-6
Molekylær formel C29 H32 N2 O3
Molekylvægt (g/mol) 456.58
Synonym 2-(4-hydroxyphenyl)-3-methyl-1-[[4-[2-(1-piperidinyl)ethoxy]phenyl]methyl]-1H-indol-5-ol, ERA 923
IUPAC navn 2-(4-hydroxyphenyl)-3-methyl-1-[[4-(2-piperidin-1-ylethoxy)phenyl]methyl]indol-5-ol
SMIL Cc1c(c2ccc(O)cc2)n(Cc3ccc(OCCN4CCCCC4)cc3)c5ccc(O)cc15
Tekniske data
Kemisk navn eller materiale Pipendoxifene
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C29H32N2O3/c1-21-27-19-25(33)11-14-28(27)31(29(21)23-7-9-24(32)10-8-23)20-22-5-12-26(13-6-22)34-18-17-30-15-3-2-4-16-30/h5-14,19,32-33H,2-4,15-18,20H2,1H3
Formel vægt 456.2413
Procent renhed >95%