Få mere at vide
Pipazetate Hydrochloride, Mikromol™
Beskrivelse
Pipazetate Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Pipazetate Hydrochloride |
| CAS | 11-7-6056 |
| Analyt- eller komponentnavne | Pipazetate Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C21 H25 N3 O3 S . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 10H-Pyrido[3,2-b][1,4]benzothiazine-10-carboxylic acid, 2-(2-piperidinoethoxy)ethyl ester, monohydrochloride (8CI), 10H-Pyrido[3,2-b][1,4]benzothiazine-10-carboxylic acid, 2-[2-(1-piperidinyl)ethoxy]ethyl ester, monohydrochloride (9CI), Ethanol, 2-(2-piperidinoethoxy)-, 10H-pyrido[3,2-b][1,4]benzothiazine-10-carboxylate (ester), monohydrochloride (8CI), 2-(2-Piperidinoethoxy)ethyl 10H-pyrido[3,2-b][1,4]benzothiazine-10-carboxylate monohydrochloride, Lenopect, Pipazetate hydrochloride, Pipazethate hydrochloride, Pipazethate monohydrochloride, Selgon, Selvigon, Selvjgon, Theratuss, Theratuss hydrochloride, 10H-Pyrido[3,2-b][1,4]benzothiazine-10-carboxylic acid, 2-[2-(1-piperidinyl)ethoxy]ethyl ester, hydrochloride (1:1), Pipazetate Hydrochloride |
| InChI formel | InChI=1S/C21H25N3O3S.ClH/c25-21(27-16-15-26-14-13-23-11-4-1-5-12-23)24-17-7-2-3-8-18(17)28-19-9-6-10-22-20(19)24;/h2-3,6-10H,1,4-5,11-16H2;1H |
| SMIL | Cl.O=C(OCCOCCN1CCCCC1)N2c3ccccc3Sc4cccnc24 |
| IUPAC navn | 2-(2-piperidin-1-ylethoxy)ethyl pyrido[3,2-b][1,4]benzothiazine-10-carboxylate;hydrochloride |
| Vis mere |