Få mere at vide
Pioglitazone-d4 (major), TRC
Artikelnummer. 30426868
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426868 1 mg
30415429 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426868 Leverandør Toronto Research Chemicals Leverandørnr. TRCP4710021MG
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pioglitazone-d4
CAS 1134163-31-7
Renhedsgrad noter HPLC
Molekylær formel C19H16D4N2O3S
Mængde 1 mg
Synonym 5-[[4-[2-(5-Ethyl-2-pyridinyl]ethoxy-d4]phenyl]methyl]-2,4-thiazolidinedione, AD-4833-d4, Actos-d4, U-72107A-d4
InChI formel InChI=1S/C19H20N2O3S/c1-2-13-3-6-15(20-12-13)9-10-24-16-7-4-14(5-8-16)11-17-18(22)21-19(23)25-17/h3-8,12,17H,2,9-11H2,1H3,(H,21,22,23)/i9D2,10D2
SMIL CCC1=CC=C(C([2H])([2H])C([2H])([2H])OC2=CC=C(CC3C(NC(S3)=O)=O)C=C2)N=C1
Molekylvægt (g/mol) 360.46
Formel vægt 360.1446
Procent renhed >95