missing translation for 'onlineSavingsMsg'
Learn More
Pinoresinol Diglucoside, MedChemExpress Product Code.: 16481468

Pinoresinol Diglucoside, MedChemExpress

Product Code. p-200101475
5 mg, 5mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16481468

Brand: Medchem Express HYN0657

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pinoresinol Diglucoside is one of the major lignans with various pharmacological activities which could be isolated from Duzhong and other plant species.

Pinoresinol Diglucoside is one of the major lignans with various pharmacological activities which could be isolated from Duzhong and other plant species.

Specifications

Chemical Name or Material Pinoresinol Diglucoside
CAS 63902-38-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C32H42O16
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (146.48 mM; Need ultrasonic)
SMILES COC(C=C1[C@@H]2[C@](CO[C@@H]3C4=CC(OC)=C(O[C@@H]([C@@H]([C@@H](O)[C@@H]5O)O)O[C@@H]5CO)C=C4)([H])[C@]3([H])CO2)=C(C=C1)O[C@@H]([C@@H]([C@@H](O)[C@@H]6O)O)O[C@@H]6CO
Molecular Weight (g/mol) 682.67
Formula Weight 682.67
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Pinoresinol Diglucoside, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.