missing translation for 'onlineSavingsMsg'
Få mere at vide

Pinacidil monohydrate, MedChemExpress

Artikelnummer. 16436028
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
Pakningsstørrelse:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16436028 10 mg -
16473416 25 mg -
16428637 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16436028 Leverandør Medchem Express Leverandørnr. HY14290A

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Pinacidil (P-1134) monohydrate, an antihypertensive drug, is a potassium channel activator.

Pinacidil (P-1134) monohydrate, an antihypertensive drug, is a potassium channel activator.

Tekniske data

Kemisk navn eller materiale Pinacidil monohydrate
CAS 85371-64-8
Farve White
Renhedsgrad noter Research
Molekylær formel C13H21N5O
Mængde 10 mg
Synonym P-1134 monohydrate
Opløselighedsinformation DMSO : 100 mg/mL (379.74 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (189.87 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMIL [H]O[H].CC(C)(C)C(N/C(NC#N)=N/C1=CC=NC=C1)C
Molekylvægt (g/mol) 263.34
Formel vægt 263.34
Procent renhed 95.0%
Grad Research
Fysisk form Powder
Vis mere Vis mindre

Research purposes only

Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.