Få mere at vide
Pimelic Acid-d4, TRC
Artikelnummer. 30418957
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30418957 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418957 Leverandør Toronto Research Chemicals Leverandørnr. TRCP44504210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,5-Pentanedicarboxylic-1,1,5,5-d4 acid
CAS 19031-56-2
Molekylær formel C7 2H4 H8 O4
Mængde 10 mg
Synonym Heptanedioic-2,2,6,6-d4 acid (9CI), Pimelic-2,2,6,6-d4 acid (8CI), 1,5-Pentanedicarboxylic-1,1,5,5-D4 acid, Pimelic acid-d4
InChI formel InChI=1S/C7H12O4/c8-6(9)4-2-1-3-5-7(10)11/h1-5H2,(H,8,9)(H,10,11)/i4D2,5D2
SMIL [2H]C([2H])(CCCC([2H])([2H])C(=O)O)C(=O)O
IUPAC navn 2,2,6,6-tetradeuterioheptanedioic acid
Molekylvægt (g/mol) 164.19
Formel vægt 164.0987