Learn More
Invitrogen™ PIM2 Recombinant Human Protein
PIM2 is a proto-oncogene.
Brand: Invitrogen™ PR6347B
Additional Details : Weight : 0.01000kg
Description
PIM2 phosphorylates BAD, which partially inhibits its apopotopic function.
Specifications
NP_006866 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
Pim2 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
63.7kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
PIM Kinase Family | |
Recombinant | |
Baculovirus |