Få mere at vide
Pilsicainide Hydrochloride, TRC
Artikelnummer. 30428122
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428122 10 mg
30414266 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30428122 Leverandør Toronto Research Chemicals Leverandørnr. TRCP44150810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pilsicainide Hydrochloride
CAS 88069-49-2
Molekylær formel C17H24N2O . HCl
Mængde 10 mg
Synonym SUN 1165, Sunrythm, N-(2,6-Dimethylphenyl)tetrahydro-1H-pyrrolizine-7a(5H)acetamide Monohydrochloride
InChI formel InChI=1S/C17H24N2O.ClH/c1-13-6-3-7-14(2)16(13)18-15(20)12-17-8-4-10-19(17)11-5-9-17;/h3,6-7H,4-5,8-12H2,1-2H3,(H,18,20);1H
SMIL Cl.Cc1cccc(C)c1NC(=O)CC23CCCN2CCC3
IUPAC navn N-(2,6-dimethylphenyl)-2-(1,2,3,5,6,7-hexahydropyrrolizin-8-yl)acetamide;hydrochloride
Molekylvægt (g/mol) 308.8
Formel vægt 308.1655