missing translation for 'onlineSavingsMsg'
Få mere at vide

Abnova™ PILRA Recombinant Protein

Artikelnummer. 16179282
Klik for at se tilgængelige muligheder
Mængde:
10 μg
25 μg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 16179282

Brand: Abnova™ H00029992P01.10ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Human PILRA full-length ORF ( AAH17812, 1 a.a. - 226 a.a.) recombinant protein with GST-tag at N-terminal

Cell signaling pathways rely on a dynamic interaction between activating and inhibiting processes. SHP-1-mediated dephosphorylation of protein tyrosine residues is central to the regulation of several cell signaling pathways. Two types of inhibitory receptor superfamily members are immunoreceptor tyrosine-based inhibitory motif (ITIM)-bearing receptors and their non-ITIM-bearing, activating counterparts. Control of cell signaling via SHP-1 is thought to occur through a balance between PILRalpha-mediated inhibition and PILRbeta-mediated activation. These paired immunoglobulin-like receptor genes are located in a tandem head-to-tail orientation on chromosome 7. This particular gene encodes the ITIM-bearing member of the receptor pair, which functions in the inhibitory role. Alternative splicing has been observed at this locus and three variants, each encoding a distinct isoform, are described. (provided by RefSeq)

  • Molecular weight: 50.6kDa
  • Preparation method: in vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
  • Quality Control Testing:12.5% SDS-PAGE stained with Coomassie Blue

Best use within three months from the date of receipt of this protein

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Tekniske data

Adgangsnummer AAH17812
Til brug med (applikation) Antibody Production, Protein Array, ELISA, Western Blot
Formulering 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gen-id (Entrez) 29992
Molekylvægt (g/mol) 51
Navn PILRA (Human) Recombinant Protein (P01)
pH-område 8
Fremstillingsmetode In vitro wheat germ expression system
Oprensningsmetode Glutathione Sepharose 4 Fast Flow
Kvalitetskontrol test 12.5% SDS-PAGE Stained with Coomassie Blue.
Mængde 10 μg
Kilde Wheat Germ (in vitro)
Immunogen MGRPLLLPLLPLLLPPAFLQPSGSTGSGPSYLYGVTQPKHLSASMGGSVEIPFSFYYPWELATAPDVRISWRRGHFHGQSFYSTRPPSIHKDYVNRLFLNWTEGQKSGFLRISNLQKQDQSVYFCRVELDTRSSGRQQWQSIEGTKLSITQGQQRTKATTPAREPFQNTEEPYENIRNEGQNTDPKLNPKLHLTQSTSQPPSPQEPPERDPVLCLKGLTNGQPSQD
Opbevaringskrav Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias FDF03
Fælles navn PILRA
Gen symbol PILRA
Krydsreaktivitet Human
Arter Wheat Germ (in vitro)
Rekombinant Recombinant
Protein tag GST
Form Solution
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.