missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Human PIK3CB/PIK3R2 (p110 beta/p85 beta), His Tag Recombinant Protein

Artikelnummer. 17188805
Klik for at se tilgængelige muligheder
Mængde:
1 mg
100 μg
20 μg
Pakningsstørrelse:
1mg
20µg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 17188805

Brand: Invitrogen™ A33395

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This PIK3CB/PIK3R2 (p110 beta/p85 beta) recombinant human protein (full length) was expressed in insect cells. PIK3CB/PIK3R2 (p110 beta/p85 beta) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for PIK3CB is NP_006210.1 and for PIK3R2 is NP_852664.1.

PIK3CB/PIK3R2 lipid kinase (p110 beta/p85 beta) or Phosphatidylinositol 3-kinase (PI3K) phosphorylate the 3-prime OH position of the inositol ring of inositol lipids which functions in signaling pathways that regulate cell growth by virtue of their activation in response to various mitogenic stimuli. PI3Ks are composed of a 110-kDa catalytic subunit and a 85-kDa adaptor subunit. PIK3CB plays a critical role in regulating the formation and stability of alpha-2B-beta-3 integrin adhesion bonds, which are necessary for shear force-induced platelet activation. PIK3CB sustains alpha-2B-beta-3 integrin activation and stabilized platelet aggregation by regulating both integrin-dependent calcium flux and Gi activation of RAP1B.
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer O00459, P42338
Koncentration See Label
Til brug med (applikation) Kinase Assay
Formulering 50mM sodium phosphate with 0.25mM DTT, 150mM Imidazole, 25% glycerol, 0.1mM PMSF, 300mM NaCl and no preservative; pH 7
Gen-id (Entrez) 5291, 5296
Molekylvægt (g/mol) 123.7-83.5 kDa
Navn Human PIK3CB/PIK3R2 (p110 beta/p85 beta), His Tag
pH-område 7
Oprensningsmetode Purified
Mængde 1 mg
Opbevaringskrav -80°C, Avoid Freeze/Thaw Cycles
Regulatorisk status RUO
Gene Alias PIK3CB; PIK3R2
Fælles navn PIK3CB/PIK3R2
Gen symbol PIK3CB, PIK3R2
KinaseFamily PI3 Kinase
Produkttype Protein
Konjugeret Unconjugated
Arter Human
Rekombinant Recombinant
Protein tag His-tag
Sekvens Full length
Indhold og opbevaring -80°C, Avoid Freeze/Thaw Cycles
Forsendelsestilstand Dry Ice
Udtrykssystem Insect cells
Protein form Full Length, Recombinant, Active
Form Liquid
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.