missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIK3C2A (PI3K-C2 alpha) Recombinant Human Protein
The Class II PI3 kinases, including PIK3C2A, phosphorylate the 3 position of PI or PI4P.
13290.00 DKK - 57500.00 DKK
Specifications
Accession Number | NP_002636.1 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Molecular Weight (g/mol) | 171.1kDa |
Storage Requirements | Kinase proteins are shipped on dry ice and should be stored at -80°C. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15488703
|
Invitrogen™
PR9053A |
100μg |
13290.00 DKK
100µg |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15408793
|
Invitrogen™
PV5587 |
1mg |
57500.00 DKK
1mg |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Though not as well studied as the Class I PI3 kinases, the Class II enyzmes have been linked to diverse, recpetor-mediated cellular processes including insulin signalling, neuronal survival, and growth factor signalling.
Specifications
NP_002636.1 | |
171.1kDa | |
RUO - research use only | |
PIK3C2A (PI3K-C2 alpha) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Catalytic | |
PI3 Kinase Family | |
Recombinant | |
Baculovirus |