missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ PIK 90
GREENER_CHOICE

Artikelnummer. 30000011
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30000011 10 mg
30000036 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30000011 Leverandør Tocris Bioscience™ Leverandørnr. 7902/10

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Potent PI3K inhibitor, also used in hPSCs mesendoderm differentiation

PIK 90 is a potent inhibitor of PI 3-K α/γ/δ kinases (IC50 = 11, 18, 47, 58 and 64 nM against p110α, p110γ, PI 3-KC2α, p110δ and PI 3-KC2α, respectively) as well as PIKKs (IC50 = 13 nM against DNA-PK). It inhibits the fMLP-stimulated phosphorylation of Akt and impairs polarity and chemotaxis in dHL60 cells. It suppresses insulin-stimulated phosphorylation of Akt and rpS6 in 3T3-L1 adipocytes and L6 myotubes in vitro and prevents insulin-induced blood glucose reduction in mice in vivo. PIK 90 can be used in combination with other small molecules and proteins for mesendoderm induction in protocols to differentiate iPSCs into organ-specific mesenchyme.
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale N-(2,3-Dihydro-7,8-dimethoxyimidazo[1,2-c]quinazolin-5-yl)-3-pyridinecarboxamide
CAS 677338-12-4
Mængde 10 mg
Mål PI 3-Kinase Inhibitors
Molekylær formel C18H17N5O3
Renhed 0.98
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.