missing translation for 'onlineSavingsMsg'
Få mere at vide

Pigment Yellow 42 (Technical Grade), TRC

Artikelnummer. 30010231
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010231 10 g
30010626 100 g
30003272 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30010231 Leverandør Toronto Research Chemicals Leverandørnr. TRCP66826010G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 51274-00-1
Molekylær formel Fe2 O3
Molekylvægt (g/mol) 159.6882
Synonym 362R, AC 2544, AZ 138, Anchor FY 766, Ariabel Yellow 300407, BTY 101W, Bayferrox 3290, Burnt Sienna TY, C 7055, C.I.77492, CI 77492 yellow, CM 3F30TRY, CM 3FA70ERH, CTD 7201, CWD 8942, Cameleon Yellow, Cappoxyt 4214X, Cappoxyt Yellow 4214, CappoxytYellow 4214C, Cappoxyt Yellow 4214X, Chromaflo 888-1810, Colanyl Oxide Yellow R 132CN, Colanyl R 132CN, Cosmetic Yellow, Creasperse YellowCP 080, Disperse HG 457, Dittany Y 77492, E 172 Yellow, EC 481, EMF Color Ochre HL, Ecosperse Oxide Yellow RA 100CN, FZ 1000, Ferric oxide,yellow, G 131M, G 313M, GA 7311, GEH 102, Goethite yellow, HP 6042, Iron Oxide Orange Transparent 188VN, Iron Oxide Yellow 214501, Iron OxideYellow 301, Iron Oxide Yellow 311, Iron Oxide Yellow 313, Iron Oxide Yellow 420, Iron Oxide Yellow Transp. 088VN, Iron Yellow, Iron Yellow 313, IronYellow AZ 138, Iron Yellow S 313, Iron hydroxide oxide yellow, Iron oxide yellow, KH 70401, Kobo Yellow WSJ 20EYAMP, L 1, L 1 (pigment), L 1916, L1918, LB 100E172, LL 100PD, LL-XLO, Luconyl Yellow 1916, MF 5050 Yellow, Mapico Yellow 1050, Mapico Yellow 5, Mapico Yellow LL-XLO, MapicoYellow Lemon, NX 512 Yellow, Oxide Yellow 3910, PS 86-1347, PW 895, PY 42, Pigment Yellow 42, Pure Yellow Oxide YO 6087, Puricolor YellowPYE 42, R 131CN, R 132CN, R 2087, S 313, S 920, S 920 (pigment), S 920M, S 960, S 960 (pigment), SZ 7499, Sicoflush L Yellow 1916, Sicoflush LYellow 1916C4, Sicoflush P Yellow 1916, Sicopharm Yellow, Sicotrans, Sicovit Yellow 10, Sicovit Yellow 10E172, SunCROMA Yellow Iron Oxide, SunPURO Yellow, Tarox Yellow Lemon, Timbasol PW 895, Toda Color Y 2, Toda Color Yellow 48, Toda Yellow 48, Trans Oxide Yellow AC 2544, Transparent YellowOxide 1852, Unipure LC 182, Unipure Yellow LC 181, Unipure Yellow LC 182, Unipure Yellow LC 182EM, Unisperse Yellow Oxide M-S, WD-TIOY30E, WS-Y 042, Xeracolor Yellow Oxide, Xeracolour Yellow Oxide, Xfast Yellow 1916
IUPAC navn oxo(oxoferriooxy)iron
SMIL O=[Fe]O[Fe]=O

Tekniske data

Kemisk navn eller materiale Pigment Yellow 42
Mængde 10 g
InChI formel InChI=1S/2Fe.3O
Formel vægt 159.8546
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.