missing translation for 'onlineSavingsMsg'
Få mere at vide

Pigment Violet 23, Technical grade, TRC

Artikelnummer. 30426544
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426544 1 g
30413339 250 mg
30410858 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30426544 Leverandør Toronto Research Chemicals Leverandørnr. TRCP4378181G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 215247-95-3
Molekylær formel C34H22Cl2N4O2
Molekylvægt (g/mol) 589.47
Synonym C.I. Pigment Violet 23 (8CI), 9,19-Dichloro-5,15-diethyl-5,15-dihydrodiindolo[2,3-c:2',3'-n]triphenodioxazine, C.I. 51319, CFP-FF 802V, Carbazole Dioxazine Violet, Carbazole Violet, Carbazole Violet 23, Chromofine Violet 6510PK, Chromofine Violet RE, Chromofine Violet RZS, Cosmenyl Violet RL, Creanova 877-8895, Cromophtal Violet GT, Cyanadur Violet, Dioxazine Violet, Dioxazine purple, EB Violet 4B7906, EMC Violet RL 10, Fast Violet RL, Fast Violet RL 3R Base, Fastogen Super Violet 140V, Fastogen Super Violet LBP 02, Fastogen Super Violet RN, Fastogen Super Violet RN-S, Fastogen Super Violet RN-SU 02, Fastogen Super Violet RTS, Fastogen Super Violet RVS, Fastogen Super Violet RXE, Fastogen Super Violet RXS, Fastogen Super Violet RZS, Flexiverse Violet 23, Fuji Fast Violet 23, Heliofast Red Violet EE, Heliofast Violet BN, Heliogen Violet, Heliogen Violet R Toner, Hostafine Violet RL, Hostaperm RL-NF, Hostaperm Violet BL, Hostaperm Violet BL 01, Hostaperm Violet P-RL, Hostaperm Violet RL, Hostaperm Violet RL 02, Hostaperm Violet RL Special, Hostaperm Violet RL Special 14-4007, Hostaperm Violet RL Special TS, Hostaperm Violet RL-COF, Hostaperm Violet RL-COF 02, Hostaperm Violet RL-COF-VP 2777, Hostaperm Violet RL-ED-LV 3064, Hostaperm Violet RL-NF, Hostaperm Violet RL-SP, Hostaperm Violet RL-SPL, IJX 1880, Indofast Violet 23, Lake Fast Violet RL, Lake Fast Violet RLB, Lionogen Violet FG 6141G, Lionogen Violet HR, Lionogen Violet R 6100, Lionogen Violet R 6200, Lionogen Violet RL, Lionol Violet HR, Luconyl Violet 5894, Monolite Fast Violet R, Novotex Violet BL VP 2435, Novotex Violet BL-PC VP 2429, Orcobrite 4BN, Orcobrite Pigment Violet 4BN, Oriental Fast Violet BL, PV 23, PV Fast Violet BL, PV Fast Violet BLP, PV Fast Violet RL, PV Fast Violet RL-SPE, Paliogen Violet 5890, Paliogen Violet L 5890, Permanent Violet, Permanent Violet R, Permanent Violet RL, Pigment Fast Violet RL, Pigment Violet 23, RL-COF 02LV3101, Sandorin Violet BL, Sanyo Permanent Violet BL-D 422, Sumikacoat Fast Violet RSB, Sumitomo Fast Violet RL 3R Base, Sumitomo Fast Violet RL Base, Sumitone Fast Violet RL, Sumitone Fast Violet RL 4R, Sumitone Fast Violet RL 4R Base, Sumitone Fast Violet RL Base, Sumitone Fast Violet RLS, Sun Violet VHD 6003, Sunfast Violet 23, Symuler Fast Violet BBL, Symuler Fast Violet BBLN, Unisperse Violet B-E, Unisperse Violet B-S, VHD 6003, Violet A-RL 100, Violet E-BL, Violet P-RL, Violet RL-NF, Viscofil Violet BLN, Vynamon Violet 2B
SMIL CCn1c2ccccc2c3c4OC5=C(Cl)C6=Nc7ccc8c(c7OC6=C(Cl)C5=Nc4ccc13)c9ccccc9n8CC

Tekniske data

Kemisk navn eller materiale Pigment Violet 23, Technical grade
Mængde 1 g
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.