Få mere at vide
Pigment Red 170, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Pigment Red 170 |
| CAS | 2786-76-7 |
| Molekylær formel | C26 H22 N4 O4 |
| Mængde | 5 g |
| Synonym | 2-Naphthalenecarboxamide','4-[[4-(aminocarbonyl)phenyl]azo]-N-(2-ethoxyphenyl)-3-hydroxy- (9CI), 2-Naphtho-o-phenetidide','4-[(p-carbamoylphenyl)azo]-3-hydroxy- (7CI,8CI), 4-[2-[4-(Aminocarbonyl)phenyl]diazenyl]-N-(2-ethoxyphenyl)-3-hydroxy-2-naphthalenecarboxamide, 4708 Permanent Red F 5RK, Brilliant Carmine 7010, C.I. 12475, C.I. PR 170, C.I. Pigment Red 170, Eljon Red NRO, Euderm Red, F 2RK, F 3RK70, Fuji Fast Red 2200, Graphtol Red F 3RK70, Graphtol Red F3RK 70-CN09, Graphtol Red NFB, Hostaperm Red F 5RK, JHR 1703L, KP Plast P-Red 3RH, L 1/8 Red F 3RK70, Lysopac Red 7030C, Naphthol AS Red, Naphthol Red 170, Naphthol Red 3172, Naphthol Red 5R, Naphtol Red B 7032C, Novoperm Red F 2RK70, Novoperm Red F 2RK70N, Novoperm Red F 3RK, Novoperm Red F 3RK70, Novoperm Red F 5RK, Novoperm Red F 6RK, PR 170, Permanent Red BH 5RK, Permanent Red F 3RK, Permanent Red F 3RK70, Permanent Red F 5RK, Permanent Red P-FK, Pigment Red 170, Red F 3RK70, Red F 5RK, Sanyo Brilliant Carmine 7009, Seikafast Red 3820, Seikafast Red 3891, Suimei Fast Red ER, Sumitone Fast Red BRS, Sunbrite Red 170 |
| InChI formel | InChI=1S/C26H22N4O4/c1-2-34-22-10-6-5-9-21(22)28-26(33)20-15-17-7-3-4-8-19(17)23(24(20)31)30-29-18-13-11-16(12-14-18)25(27)32/h3-15,31H,2H2,1H3,(H2,27,32)(H,28,33) |
| SMIL | CCOc1ccccc1NC(=O)c2cc3ccccc3c(N=Nc4ccc(cc4)C(=O)N)c2O |
| IUPAC navn | 4-[(E)-(4-carbamoylphenyl)diazenyl]-N-(2-ethoxyphenyl)-3-hydroxynaphthalene-2-carboxamide |
| Molekylvægt (g/mol) | 454.477 |
| Formel vægt | 454.164 |