Få mere at vide
Pigment Red 149 (Technical Grade), TRC
Artikelnummer. 30424837
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424837 500 mg
30422676 250 mg
30410412 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30424837 Leverandør Toronto Research Chemicals Leverandørnr. TRCP437825500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4948-15-6
Molekylær formel C40H26N2O4
Molekylvægt (g/mol) 598.65
Synonym Anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetrone, 2,9-bis(3,5-dimethylphenyl)-, 3,4,9,10-Perylenetetracarboxylic 3,4:9,10-diimide, N,N'-di-3,5-xylyl- (6CI,7CI,8CI), 2,9-Bis(3,5-dimethylphenyl)anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetrone, C.I. 71137, C.I. PR 149, C.I. Pigment Red 149, Dymic MBR 155, Exalite 613, Kayaset Red E-GR, Lionogen Red H 103B, MBR 155, N,N’-Bis(3,5-dimethylphenyl)-3,4:9,10-perylenebis(dicarboximide), N,N’-Bis(3,5-dimethylphenyl)-3,4:9,10-perylenebis(dicarboxyimide), N,N’-Bis(3,5-dimethylphenyl)perylene-3,4,9,10-tetracarboxylic 3,4:9,10-diimide, N,N’-Bis(3,5-xylyl)perylene-3,4:9,10-bis(dicarboximide), N,N’-Bis(3',5'-dimethylphenyl)perylene-3,4,9,10-dicarboimide, N,N’-Di(3,5-xylyl)perylene-3,4:9,10-bis(dicarboximide), Novoperm Red BL, PR 149, PR 149 Red, PV Fast Red B, PV Solid Red B, Palamid Red 35-8005, Paliogen Red K 3580, Permanent Red BC, Permanent Red BL, Perylene PV Fast Red B, Perylene-3,4,9,10-tetracarboxylic acid bis(3',5'-dimethylanilide), Red 04, Red K 3580
SMIL Cc1cc(C)cc(c1)N2C(=O)c3ccc4c5ccc6C(=O)N(C(=O)c7ccc(c8ccc(C2=O)c3c48)c5c67)c9cc(C)cc(C)c9
Tekniske data
Kemisk navn eller materiale Pigment Red 149 (Technical Grade)
Mængde 500 mg
InChI formel InChI=1S/C40H26N2O4/c1-19-13-20(2)16-23(15-19)41-37(43)29-9-5-25-27-7-11-31-36-32(40(46)42(39(31)45)24-17-21(3)14-22(4)18-24)12-8-28(34(27)36)26-6-10-30(38(41)44)35(29)33(25)26/h5-18H,1-4H3
Formel vægt 598.1893