Få mere at vide
Pigment Red 112 (tech grade), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Pigment Red 112 |
| CAS | 6535-46-2 |
| Molekylær formel | C24 H16 Cl3 N3 O2 |
| Mængde | 100 mg |
| Synonym | 2-Naphthalenecarboxamide','3-hydroxy-N-(2-methylphenyl)-4-[(2,4,5-trichlorophenyl)azo]- (9CI), C.I. Pigment Red 112 (7CI,8CI), 3-Hydroxy-N-(2-methylphenyl)-4-[2-(2,4,5-trichlorophenyl)diazenyl]-2-naphthalenecarboxamide, C.I. 12370, C.I. PR 112, Colanyl Red FGRG 100, Colanyl Red FGRX, Dispers Red 38-5507, Disperse Red 38-5507, Disperse Red SDP 110T, Fast Red FGR, Flexonyl Red FGR-LA, Helio Fast Red BB, Helio Fast Red BBN, Helio Fast Red BBT, Hostafine Red FGR, Irgalite Red 3RS, Isol Aryl Red GR, KET Red 304, Luconyl Red 3855, Naphthol Red FGR, Oriental Fast Red GR, PR 112, Permanent Red FGR, Permanent Red FGR 02, Permanent Red FGR 70, Permanent Red GY, Permanent Red L, Pigment Red 112, Renol Red FGR-HW, Sanyo Permanent Red 1206, Sanyo Permanent Red FN, Sanyo Permanent Red FNG, Segnale Light Red FGR, Sico Fast Red L 3855, SicoFast Red 3855, Special Red FGR, Unisperse Red 3RS-E2, Unisperse Red 3RS-P, Xfast Red 3855 |
| InChI formel | InChI=1S/C24H16Cl3N3O2/c1-13-6-2-5-9-20(13)28-24(32)16-10-14-7-3-4-8-15(14)22(23(16)31)30-29-21-12-18(26)17(25)11-19(21)27/h2-12,31H,1H3,(H,28,32) |
| SMIL | Cc1ccccc1NC(=O)c2cc3ccccc3c(N=Nc4cc(Cl)c(Cl)cc4Cl)c2O |
| IUPAC navn | 3-hydroxy-N-(2-methylphenyl)-4-[(E)-(2,4,5-trichlorophenyl)diazenyl]naphthalene-2-carboxamide |
| Molekylvægt (g/mol) | 484.76 |
| Formel vægt | 483.0308 |