Få mere at vide
Pigment Orange 13 (Technical Grade), TRC
Artikelnummer. 30427209
Skift visning
Mængde:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427209 5 g
30410886 50 g
30410729 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30427209 Leverandør Toronto Research Chemicals Leverandørnr. TRCP4378135G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3520-72-7
Molekylær formel C32H24Cl2N8O2
Molekylvægt (g/mol) 623.49
Synonym 4,4'-[(3,3'-Dichloro[1,1'-biphenyl]-4,4'-diyl)bis(2,1-diazenediyl)]bis[2,4-dihydro-5-methyl-2-phenyl-3H-pyrazol-3-one], Atul Vulcan Fast Pigment Orange G, Benzidine Orange 45-2850, Benzidine Orange 45-2880, Benzidine Orange Toner, Benzidine Orange WD 265, C.I. 21110, Calcotone Orange R, Carnelio Orange G, DCC 1802Dainichi Fast Orange RR, Daltolite Fast Orange G, Diarylide Orange, Disazo Orange PMP, Eljon Fast Orange G, Fast Benzidene Orange YB 3, Fast Orange J, Fastona Orange 13, Fastona Orange G, Flexonyl Orange G 100, GSR 1151, Graphtol Orange GP, Irgalite Fast Orange PG, Irgalite Orange P, Irgalite Orange PG, Irgalite Orange PX, Irgaplast Orange G, Japan Orange 204, KET Orange 501, Kromon Orange G, Latexol Fast Orange J, Lionol Orange B, Lutetia Orange J, Monolite Fast Orange G, Monolite Fast Orange GA
IUPAC navn 4-[(E)-[2-chloro-4-[3-chloro-4-[(E)-(3-methyl-5-oxo-1-phenyl-4H-pyrazol-4-yl)diazenyl]phenyl]phenyl]diazenyl]-5-methyl-2-phenyl-4H-pyrazol-3-one
SMIL CC1=NN(C(=O)C1N=Nc2ccc(cc2Cl)c3ccc(N=NC4C(=NN(C4=O)c5ccccc5)C)c(Cl)c3)c6ccccc6
Tekniske data
Kemisk navn eller materiale Pigment Orange 13 (Technical Grade)
Mængde 5 g
InChI formel InChI=1S/C32H24Cl2N8O2/c1-19-29(31(43)41(39-19)23-9-5-3-6-10-23)37-35-27-15-13-21(17-25(27)33)22-14-16-28(26(34)18-22)36-38-30-20(2)40-42(32(30)44)24-11-7-4-8-12-24/h3-18,29-30H,1-2H3
Formel vægt 622.1399