Learn More
Thermo Scientific™ Pierce™ Sulfo-SANPAH, No-Weigh™ Format
Brand: Thermo Scientific™ 22589
3149.69 DKK valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Additional Details : Weight : 1.81440kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Description
Thermo Scientific™ Pierce Sulfo-SANPAH is a long-arm (18.2 Å) crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester and a photoactivatable nitrophenyl azide.
Highlights:
- Optimal photolysis occurs at 320-350nm, which limits damage to biomolecules by irradiation
- Water-soluble
- Reactive groups: sulfo-NHS ester and nitrophenyl azide (Amax= 470nm)
- Reactive toward: amino groups and various sites
Specifications
Amine-Nonselective | |
Sulfo-SANPAH | |
No | |
Upon receipt store at -20°C protected from light and moisture. | |
Ambient | |
Chemical Labeling | |
Sulfo-NHS Ester, Aryl Azide | |
Powder | |
Water | |
Yes |
By Thiols | |
492.4 | |
18.2 Å | |
No | |
Pierce™ | |
Heterobifunctional | |
Medium (10 to 30 Å) | |
50 mg | |
Standard, Single-use | |
Crosslinker |
Safety and Handling
- (SULFO-SANPAH) SULFOSUCCINIMID
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
For Research Use Only. Not for use in diagnostic procedures.