Learn More
Thermo Scientific™ Pierce™ Protein Concentrator PES, 2 to 6mL
Thermo Scientific™ Pierce™ Protein Concentrators PES are easy-to-use centrifugal devices that provide fast processing and excellent recovery of protein samples.
96.50 DKK - 902.00 DKK
Specifications
Volume (Metric) 2 to 6mL
View More Specs
Products 12
Product Code Brand MWCO Type Volume (Metric) Quantity Price Quantity & Availability  
Product Code Brand MWCO Type Volume (Metric) Quantity Price Quantity & Availability  
15522953 View Documents Thermo Scientific™
88516
10kDa Protein Concentrator 2 to 6mL 10 rxn(s) 445.00 DKK
10 reactions View alternative products
Save Up to 
»
In Stock 11
Add to basket
15471295 View Documents Thermo Scientific™
88515
3kDa Protein Concentrator 2 to 6mL 24 rxn(s) 894.00 DKK
24 reactions View alternative products
Save Up to 
»
In Stock 9
Add to basket
16341954 View Documents Thermo Scientific™
88514S
3K Spin Column, 15 mL conical 2 to 6mL 2pk 96.50 DKK
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-03-2020
Add to basket
15594279 View Documents Thermo Scientific™
88523
100K Spin Column, 15 mL conical 2 to 6mL 10pk 448.00 DKK
10 reactions View alternative products
Save Up to 
»
In Stock 4
Add to basket
15566775 View Documents Thermo Scientific™
88524
100K Spin Column, 15 mL conical 2 to 6mL 24pk 902.00 DKK
24 reactions View alternative products
Save Up to 
»
In Stock 5
Add to basket
16351954 View Documents Thermo Scientific™
88516S
10K Spin Column, 15 mL conical 2 to 6mL 2pk 96.50 DKK
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-03-2020
Add to basket
15402304 View Documents Thermo Scientific™
88522
30kDa Protein Concentrator 2 to 6mL 24 rxn(s) 894.00 DKK
24 tests View alternative products
Save Up to 
»
In Stock 14
Add to basket
15456824 View Documents Thermo Scientific™
88521
30kDa Protein Concentrator 2 to 6mL 10 rxn(s) 447.00 DKK
10 reactions View alternative products
Save Up to 
»
In Stock 5
Add to basket
16361954 View Documents Thermo Scientific™
88521S
30K Spin Column, 15 mL conical 2 to 6mL 2pk 96.50 DKK
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-03-2020
Add to basket
15431305 View Documents Thermo Scientific™
88514
3kDa Protein Concentrator 2 to 6mL 10 rxn(s) 447.00 DKK
10 reactions View alternative products
Save Up to 
»
In Stock 2
Add to basket
15432284 View Documents Thermo Scientific™
88517
10kDa Protein Concentrator 2 to 6mL 24 rxn(s) 894.00 DKK
24 tests View alternative products
Save Up to 
»
In Stock 41
Add to basket
16371954 View Documents Thermo Scientific™
88523S
100K Spin Column, 15 mL conical 2 to 6mL 2pk 96.50 DKK
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-03-2020
Add to basket
Description
Description
The Thermo Scientific™ Pierce™ concentrators are easy-to-use centrifugal devices that provide fast processing and excellent recovery of protein samples. These disposable ultrafiltration devices contain a polyethersulfone (PES) membrane for the concentration, desalting, and buffer exchange of biological samples, such as tissue culture media, antiserum, monoclonal antibody preparations, and chromatography fractions. They can also be used to remove unincorporated label following protein modification reactions. 
The PES membrane in these devices is available in five distinct molecular-weight cutoffs (MWCO) of 3K, 5K, 10K, 30K, and 100K, and can be used for processing sample volumes from 100 μL to 100 mL. The MWCOs are etched on the sides of the concentrators for easy identification, while a clear window with graduations on the side of each device allows for visual determination of the concentrated sample (retentate) volume. The unique design provides reliable and consistent results. Multiple device sizes are available to handle maximum sample volumes of 0.5 mL, 6 mL, 20 mL, and 100 mL.
  • Rapid processing—unique design minimizes membrane fouling and sample concentration of 10- to 30-fold can be achieved in 5–30 minutes for 10K MWCO (device dependent—times may vary for other MWCO’s), even with particle laden solutions.
  • High recovery—retain >90% of protein samples while removing contaminants or exchanging buffers.
  • Convenient—clear markings, wide sample chamber and removable filtrate chamber make handling simple and easy.
  • Instrument compatible—can be used with standard centrifuges utilizing either fixed-angle or swinging-bucket rotors.
Specifications
Specifications
2 to 6mL