missing translation for 'onlineSavingsMsg'
Få mere at vide

Picroside II, TRC

Artikelnummer. 30427212
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427212 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30427212 Leverandør Toronto Research Chemicals Leverandørnr. TRCP43764010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 39012-20-9
Molekylær formel C23 H28 O13
Molekylvægt (g/mol) 512.46
Synonym β-D-Glucopyranoside, (1aS,1bS,2S,5aR,6S,6aS)-1a,1b,2,5a,6,6a-hexahydro-6-[(4-hydroxy-3-methoxybenzoyl)oxy]-1a-(hydroxymethyl)oxireno[4,5]cyclopenta[1,2-c]pyran-2-yl, β-D-Glucopyranoside, 1a,1b,2,5a,6,6a-hexahydro-6-[(4-hydroxy-3-methoxybenzoyl)oxy]-1a-(hydroxymethyl)oxireno[4,5]cyclopenta[1,2-c]pyran-2-yl, [1aS-(1aα,1bβ,2β,5aβ,6β,6aα)]-, Oxireno[4,5]cyclopenta[1,2-c]pyran, β-D-glucopyranoside deriv., (1aS,1bS,2S,5aR,6S,6aS)-1a,1b,2,5a,6,6a-Hexahydro-6-[(4-hydroxy-3-methoxybenzoyl)oxy]-1a-(hydroxymethyl)oxireno[4,5]cyclopenta[1,2-c]pyran-2-yl β-D-glucopyranoside, 6-Vanilloylcatalpol, Picroside II, Vanilloyl catalpol
IUPAC navn [(1aS,1bS,2S,5aR,6S,6aS)-1a-(hydroxymethyl)-2-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2,5a,6,6a-tetrahydro-1bH-oxireno[5,6]cyclopenta[1,3-c]pyran-6-yl] 4-hydroxy-3-methoxybenzoate
SMIL COc1cc(ccc1O)C(=O)O[C@H]2[C@@H]3C=CO[C@@H](O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@@H]3[C@@]5(CO)O[C@@H]25

Tekniske data

Kemisk navn eller materiale Picroside II
Mængde 10 mg
InChI formel InChI=1S/C23H28O13/c1-31-12-6-9(2-3-11(12)26)20(30)34-18-10-4-5-32-21(14(10)23(8-25)19(18)36-23)35-22-17(29)16(28)15(27)13(7-24)33-22/h2-6,10,13-19,21-22,24-29H,7-8H2,1H3/t10-,13-,14-,15-,16+,17-,18+,19+,21+,22+,23-/m1/s1
Formel vægt 512.153
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.