Få mere at vide
Picropodophyllotoxin-d3, TRC
Artikelnummer. 30409260
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30409260 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30409260 Leverandør Toronto Research Chemicals Leverandørnr. TRCP4376221MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Picropodophyllotoxin-d3
CAS 1454782-81-0
Molekylær formel C22H19D3O8
Mængde 1 mg
Synonym Picropodophyllin-d3, AXL 1717-d3, NSC 36407-d3, [5R-(5α,5aα,8aα,9α)]-5,8,8a,9-Tetrahydro-9-hydroxy-5-(3,4,5-trimethoxy-d3-phenyl)furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, (5R,5aS,8aR,9R)-9-Hydroxy-5-(3,4,5-trimethoxy-d3-phenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one
InChI formel InChI=1S/C22H22O8/c1-25-16-4-10(5-17(26-2)21(16)27-3)18-11-6-14-15(30-9-29-14)7-12(11)20(23)13-8-28-22(24)19(13)18/h4-7,13,18-20,23H,8-9H2,1-3H3/t13-,18+,19+,20-/m0/s1/i3D3
SMIL O[C@@H]1[C@]2([H])[C@@]([H])(C(OC2)=O)[C@H](C3=CC(OC)=C(OC([2H])([2H])[2H])C(OC)=C3)C4=C1C=C(OCO5)C5=C4
Molekylvægt (g/mol) 417.42
Formel vægt 417.1503