Få mere at vide
Phytol-d5, TRC
Artikelnummer. 30422397
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30422397 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422397 Leverandør Toronto Research Chemicals Leverandørnr. TRCP398912100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phytol-d5
CAS 1361929-42-1
Molekylær formel C20 D5 H35 O
Mængde 100 mg
Synonym (7R,11R,E)-7,11,15-trimethyl-3-(methyl-d3)hexadec-2-en-4,4-d2-1-ol, (7R,11R)-7,11,15-Trimethyl-3-(methyl-d3)-2-hexadecen-4,4-d2-1-ol
InChI formel InChI=1S/C20H40O/c1-17(2)9-6-10-18(3)11-7-12-19(4)13-8-14-20(5)15-16-21/h15,17-19,21H,6-14,16H2,1-5H3/b20-15+/t18-,19-/m1/s1/i5D3,14D2
SMIL [2H]C([2H])([2H])\C(=C/CO)\C([2H])([2H])CC[C@H](C)CCC[C@H](C)CCCC(C)C
IUPAC navn (E,7R,11R)-4,4-dideuterio-7,11,15-trimethyl-3-(trideuteriomethyl)hexadec-2-en-1-ol
Molekylvægt (g/mol) 301.562
Formel vægt 301.339