Få mere at vide
Phytol-d5, TRC
Artikelnummer. 30414647
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30414647 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414647 Leverandør Toronto Research Chemicals Leverandørnr. TRCP39891210MG
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phytol-d5
CAS 1361929-42-1
Molekylær formel C20 D5 H35 O
Mængde 10 mg
Synonym (7R,11R,E)-7,11,15-trimethyl-3-(methyl-d3)hexadec-2-en-4,4-d2-1-ol, (7R,11R)-7,11,15-Trimethyl-3-(methyl-d3)-2-hexadecen-4,4-d2-1-ol
InChI formel InChI=1S/C20H40O/c1-17(2)9-6-10-18(3)11-7-12-19(4)13-8-14-20(5)15-16-21/h15,17-19,21H,6-14,16H2,1-5H3/b20-15+/t18-,19-/m1/s1/i5D3,14D2
SMIL [2H]C([2H])([2H])\C(=C/CO)\C([2H])([2H])CC[C@H](C)CCC[C@H](C)CCCC(C)C
IUPAC navn (E,7R,11R)-4,4-dideuterio-7,11,15-trimethyl-3-(trideuteriomethyl)hexadec-2-en-1-ol
Molekylvægt (g/mol) 301.562
Formel vægt 301.339