Få mere at vide
Phytanoic Acid, TRC
Artikelnummer. 30418093
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418093 100 mg
30413508 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30418093 Leverandør Toronto Research Chemicals Leverandørnr. TRCP147200100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phytanoic Acid
CAS 14721-66-5
Renhedsgrad noter HPLC
Molekylær formel C20 H40 O2
Mængde 100 mg
Synonym 3,7,11,15-Tetramethylhexadecanoic Acid, NSC 108698, Phytanic Acid
InChI formel InChI=1S/C20H40O2/c1-16(2)9-6-10-17(3)11-7-12-18(4)13-8-14-19(5)15-20(21)22/h16-19H,6-15H2,1-5H3,(H,21,22)
SMIL CC(C)CCCC(C)CCCC(C)CCCC(C)CC(=O)O
IUPAC navn 3,7,11,15-tetramethylhexadecanoic acid
Molekylvægt (g/mol) 312.53
Formel vægt 312.3028
Procent renhed >95