Learn More
Physostigmine Salicylate, Mikromol™
Product Code. 30052422
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30052422 100 mg 100mg
1 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30052422 Supplier Mikromol Supplier No. MM1707.00
 Restricted Item
Forventet forsendelse: 27-11-2026
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Physostigmine Salicylate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Eserine Salicylate
CAS 57-64-7
Analyte or Component Names Eserine Salicylate
Shipping Condition Room Temperature
Molecular Formula C15 H21 N3 O2 . C7 H6 O3
Certifications/Compliance ISO 17034
Synonym Physostigmine salicylate, (3aS,8aR)-1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethylpyrrolo[2,3-b]indol-5-yl methylcarbamate salicylate, Benzoic acid, 2-hydroxy-, compd. with (3aS,8aR)-1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethylpyrrolo[2,3-b]indol-5-yl methylcarbamate (1:1) (9CI), Benzoic acid, 2-hydroxy-, compd. with (3aS-cis)-1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethylpyrrolo[2,3-b]indol-5-yl methylcarbamate (1:1), Physostigmine salicylate (6CI), Physostigmine, monosalicylate (8CI), Pyrrolo[2,3-b]indol-5-ol, 1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethyl-, methylcarbamate (ester), (3aS,8aR)-, mono(2-hydroxybenzoate) (salt) (9CI), Pyrrolo[2,3-b]indol-5-ol, 1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethyl-, methylcarbamate (ester), (3aS-cis)-, mono(2-hydroxybenzoate) (salt), Salicylic acid, compd. with physostigmine (1:1) (8CI), (-)-Physostigmine salicylate, Eserine salicylate, Isopto Eserine
InChI Formula InChI=1S/C15H21N3O2.C7H6O3/c1-15-7-8-17(3)13(15)18(4)12-6-5-10(9-11(12)15)20-14(19)16-2;8-6-4-2-1-3-5(6)7(9)10/h5-6,9,13H,7-8H2,1-4H3,(H,16,19);1-4,8H,(H,9,10)/t13-,15+;/m1./s1
SMILES CNC(=O)Oc1ccc2N(C)[C@H]3N(C)CC[C@@]3(C)c2c1.OC(=O)c4ccccc4O
IUPAC Name [(3aR,8bS)-3,4,8b-trimethyl-2,3a-dihydro-1H-pyrrolo[2,3-b]indol-7-yl] N-methylcarbamate;2-hydroxybenzoic acid
Molecular Weight (g/mol) 413.47
Quantity 100 mg
Formula Weight 413.1951 g/mol