Learn More
Physostigmine Salicylate, LoGiCal
Product Code. 30461767
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30461767 10 mg 10mg
1 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30461767 Supplier LoGiCal Supplier No. LGCFOR1707.00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Eserine Salicylate
CAS 57-64-7
Analyte or Component Names Eserine Salicylate
Shipping Condition Room Temperature
Molecular Formula C15 H21 N3 O2 . C7 H6 O3
Certifications/Compliance ISO 9001
Synonym Physostigmine salicylate, (3aS,8aR)-1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethylpyrrolo[2,3-b]indol-5-yl methylcarbamate salicylate, Benzoic acid, 2-hydroxy-, compd. with (3aS,8aR)-1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethylpyrrolo[2,3-b]indol-5-yl methylcarbamate (1:1) (9CI), Benzoic acid, 2-hydroxy-, compd. with (3aS-cis)-1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethylpyrrolo[2,3-b]indol-5-yl methylcarbamate (1:1), Physostigmine salicylate (6CI), Physostigmine, monosalicylate (8CI), Pyrrolo[2,3-b]indol-5-ol, 1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethyl-, methylcarbamate (ester), (3aS,8aR)-, mono(2-hydroxybenzoate) (salt) (9CI), Pyrrolo[2,3-b]indol-5-ol, 1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethyl-, methylcarbamate (ester), (3aS-cis)-, mono(2-hydroxybenzoate) (salt), Salicylic acid, compd. with physostigmine (1:1) (8CI), (-)-Physostigmine salicylate, Eserine salicylate, Isopto Eserine
InChI Formula InChI=1S/C15H21N3O2.C7H6O3/c1-15-7-8-17(3)13(15)18(4)12-6-5-10(9-11(12)15)20-14(19)16-2;8-6-4-2-1-3-5(6)7(9)10/h5-6,9,13H,7-8H2,1-4H3,(H,16,19);1-4,8H,(H,9,10)/t13-,15+;/m1./s1
Solution Type Neat
SMILES CNC(=O)Oc1ccc2N(C)[C@H]3N(C)CC[C@@]3(C)c2c1.OC(=O)c4ccccc4O
IUPAC Name [(3aR,8bS)-3,4,8b-trimethyl-2,3a-dihydro-1H-pyrrolo[2,3-b]indol-7-yl] N-methylcarbamate;2-hydroxybenzoic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 413.47
Quantity 10 mg
Formula Weight 413.1951 g/mol