Learn More
uvex Phynomic AirLite A ESD Safety Glove
The Uvex phynomic airLite ESD is a light safety glove that offers a noticeable difference in wearer comfort.
463.00 DKK
Specifications
Certifications/Compliance | EN 388 (3 1 1 0 X) and DIN EN 16350:2014 |
---|---|
Coating Material | Aqua-Polymer Foam |
Color | Black |
Disposable | Reusable |
Exterior Finish | Smooth |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17767569
|
uvex
6003805 |
5 |
463.00 DKK
10 pairs |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
17777569
|
uvex
6003806 |
6 |
463.00 DKK
10 pairs |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
17787569
|
uvex
6003807 |
7 |
463.00 DKK
10 pairs |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
17797569
|
uvex
6003808 |
8 |
463.00 DKK
10 pairs |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
17707579
|
uvex
6003809 |
9 |
463.00 DKK
10 pairs |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
17717579
|
uvex
6003810 |
10 |
463.00 DKK
10 pairs |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
17727579
|
uvex
6003811 |
11 |
463.00 DKK
10 pairs |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
17737579
|
uvex
6003812 |
12 |
463.00 DKK
10 pairs |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
Description
- The lightest and most sensitive uvex phynomic cut protection glove in Cut Level A
- Outer material made from polyamide, elastane and carbon
- Breathable 'airLite aqua-polymer coating on palm and fingertips
- Safety glove with touchscreen function – for use on almost all screens, tablets and mobile phones
- Safety glove with ESD function in line with DIN EN 16350:2014 – protects workpiece against electrostatic charge
- Good grip in dry and slightly damp areas
- Free from accelerators, and health protection and skin compatibility dermatologically approved (proDERM)
- Very lightweight and breathable safety glove
- Outstanding sensitivity thanks to new breathable 'airLite' aqua-polymer coating combined with fine liner (18 gauge)
- Safety gloves with knitted cuff and ergonomic fit
Specifications
EN 388 (3 1 1 0 X) and DIN EN 16350:2014 | |
Black | |
Smooth | |
Not tested | |
Polyamide, Elastane, Carbon Fiber | |
ESD Gloves |
Aqua-Polymer Foam | |
Reusable | |
Ambidextrous | |
Yes | |
Powder Free | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
uvex Phynomic AirLite A ESD Safety Glove